Anti-PAP1 Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: PAP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
- Catalog No:
- 77819-534
- 77819-535
- Antigen Symbol:
- PAP1
- PAP1
- Antigen Name:
- Recombinant Arabidopsis thaliana Probable plastid-lipid-associated protein 1, chloroplastic protein (56-318AA)
- Recombinant Arabidopsis thaliana Probable plastid-lipid-associated protein 1, chloroplastic protein (56-318AA)
- Antigen Synonyms:
- PAP1|FBN1a|FIB1a|PGL35|At4g04020|T24H24.16|Probable plastid-lipid-associated protein 1|chloroplastic|AtPap1|Fibrillin-1a|Plastoglobulin 35|AtPGL35
- PAP1|FBN1a|FIB1a|PGL35|At4g04020|T24H24.16|Probable plastid-lipid-associated protein 1|chloroplastic|AtPap1|Fibrillin-1a|Plastoglobulin 35|AtPGL35
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- A. thaliana
- A. thaliana
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O81439
- O81439
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together