Anti-CCP110 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: CCP110
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77819-488
- 77819-487
- Antigen Symbol:
- CCP110
- CCP110
- Antigen Name:
- Centriolar coiled-coil protein of 110 kDa protein (1-280AA)
- Centriolar coiled-coil protein of 110 kDa protein (1-280AA)
- Antigen Synonyms:
- 110 kDa centrosomal protein| CCP110| Centriolar coiled coil protein 110kDa| Centrosomal protein 110 kDa| Centrosomal protein CP110| Centrosomal protein of 110 kDa| Cep110| CP110| CP110_HUMAN| DKFZp781G1416| FAN| KIAA0419
- 110 kDa centrosomal protein| CCP110| Centriolar coiled coil protein 110kDa| Centrosomal protein 110 kDa| Centrosomal protein CP110| Centrosomal protein of 110 kDa| Cep110| CP110| CP110_HUMAN| DKFZp781G1416| FAN| KIAA0419
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43303
- O43303
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together