Anti-DPP9 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DPP9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77831-905
- 77831-906
- Antigen Symbol:
- DPP9
- DPP9
- Antigen Name:
- Dipeptidyl peptidase 9 protein (289-437AA)
- Dipeptidyl peptidase 9 protein (289-437AA)
- Antigen Synonyms:
- Dipeptidyl peptidase 9|Dipeptidyl peptidase IV related protein 2|Dipeptidyl peptidase IV-related protein 2|Dipeptidyl peptidase IX|Dipeptidyl peptidase like protein 9|Dipeptidyl peptidase-like protein 9|Dipeptidylpeptidase 9|Dipeptidylpeptidase IX|DKFZp762F117|DP 9|DP9|DPLP 9|DPLP9|DPP 9|DPP IX|DPP9|DPP9_HUMAN|DPRP 2|DPRP-2|DPRP2|FLJ16073
- Dipeptidyl peptidase 9|Dipeptidyl peptidase IV related protein 2|Dipeptidyl peptidase IV-related protein 2|Dipeptidyl peptidase IX|Dipeptidyl peptidase like protein 9|Dipeptidyl peptidase-like protein 9|Dipeptidylpeptidase 9|Dipeptidylpeptidase IX|DKFZp762F117|DP 9|DP9|DPLP 9|DPLP9|DPP 9|DPP IX|DPP9|DPP9_HUMAN|DPRP 2|DPRP-2|DPRP2|FLJ16073
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q86TI2
- Q86TI2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together