Anti-bla Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: bla
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pseudomonas aeruginosa
Specifications
- Catalog No:
- 77815-298
- 77815-297
- Antigen Symbol:
- bla
- bla
- Antigen Name:
- Recombinant Pseudomonas aeruginosa Beta-lactamase OXA-10 protein (21-266AA)
- Recombinant Pseudomonas aeruginosa Beta-lactamase OXA-10 protein (21-266AA)
- Antigen Synonyms:
- bla|oxa10|pse2Beta-lactamase OXA-10|EC 3.5.2.6|Beta-lactamase PSE-2
- bla|oxa10|pse2Beta-lactamase OXA-10|EC 3.5.2.6|Beta-lactamase PSE-2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Pseudomonas aeruginosa
- Pseudomonas aeruginosa
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P14489
- P14489
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together