Anti-MYH4 Rabbit Polyclonal Antibody
77797-613
New Product
- Antigen Name:Synthesized peptide derived from the C-terminal region of MYH4.
- Antigen Symbol:MYH4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q9Y623
- Antigen Synonyms:fetal|MYH2B|Myh4|MYH4_HUMAN|MyHC-2b|MyHC-IIb|Myosin heavy chain 2b|Myosin heavy chain 4|Myosin heavy chain 4 skeletal muscle|Myosin heavy chain|Myosin heavy chain IIb|Myosin heavy chain skeletal muscle fetal|Myosin-4|skeletal muscle
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on Blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: IHC: 1:100 to 1:300, ELISA: 1:5000
Type:
Antigen: MYH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human