Anti-CYB5R1 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CYB5R1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77841-425
- 77841-424
- Antigen Symbol:
- CYB5R1
- CYB5R1
- Antigen Name:
- NADH-cytochrome b5 reductase 1 protein (29-305AA)
- NADH-cytochrome b5 reductase 1 protein (29-305AA)
- Antigen Synonyms:
- b5R.1|B5R1|B5R2|CYB5R1|Cytochrome b5 reductase 1|EC 1.6.2.2|Humb5R2|NAD|NAD(P)H:quinone oxidoreductase type 3 polypeptide A2|NAD(P)H:quinone oxidoreductase type 3, polypeptide A2|NADH cytochrome b5 reductase 1|NADH-cytochrome b5 reductase 1|NB5R1_HUMAN|NQO3A2|UNQ3049/PRO9865
- b5R.1|B5R1|B5R2|CYB5R1|Cytochrome b5 reductase 1|EC 1.6.2.2|Humb5R2|NAD|NAD(P)H:quinone oxidoreductase type 3 polypeptide A2|NAD(P)H:quinone oxidoreductase type 3, polypeptide A2|NADH cytochrome b5 reductase 1|NADH-cytochrome b5 reductase 1|NB5R1_HUMAN|NQO3A2|UNQ3049/PRO9865
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UHQ9
- Q9UHQ9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together