Anti-PHGDH Rabbit Recombinant Polyclonal Antibody (FITC)
New Product
Type:
Antigen: PHGDH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77806-609
- 77806-610
- Antigen Symbol:
- PHGDH
- PHGDH
- Antigen Name:
- D-3-phosphoglycerate dehydrogenase protein (2-251AA)
- D-3-phosphoglycerate dehydrogenase protein (2-251AA)
- Antigen Synonyms:
- 3 PGDH|3-PGDH|3-phosphoglycerate dehydrogenase|3PGDH|D-3-phosphoglycerate dehydrogenase|EC 1.1.1.95|Epididymis secretory protein Li 113|HEL S 113|NLS|NLS1|PDG|PGAD|PGD|PGDH|PGDH3|Phgdh|PHGDHD|Phosphoglycerate dehydrogenase|SERA|SERA_HUMAN
- 3 PGDH|3-PGDH|3-phosphoglycerate dehydrogenase|3PGDH|D-3-phosphoglycerate dehydrogenase|EC 1.1.1.95|Epididymis secretory protein Li 113|HEL S 113|NLS|NLS1|PDG|PGAD|PGD|PGDH|PGDH3|Phgdh|PHGDHD|Phosphoglycerate dehydrogenase|SERA|SERA_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O43175
- O43175
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together