Anti-TAS1R3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77830-518
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Taste receptor type 1 member 3 protein (400-570AA)
- Antigen Symbol:TAS1R3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q7RTX0
- Antigen Synonyms:G protein coupled receptor|Sac|Saccharin preference|Sweet taste receptor T1R3|T1 R3|T1R 3|T1R3|TAS1 R3|TAS1R 3|TAS1R3|Taste receptor type 1 member 3|Taste receptor type 1 member 3 precursor|TR 3|TR3|TS1R3_HUMAN
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: TAS1R3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human