Anti-PCIF1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PCIF1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77842-054
- 77842-055
- Antigen Symbol:
- PCIF1
- PCIF1
- Antigen Name:
- Phosphorylated CTD-interacting factor 1 protein (201-500AA)
- Phosphorylated CTD-interacting factor 1 protein (201-500AA)
- Antigen Synonyms:
- bA465L10.1|C20orf67|Pcif1|PCIF1_HUMAN|PDX1 C terminal inhibiting factor 1|Phosphorylated CTD interacting factor 1|Phosphorylated CTD-interacting factor 1
- bA465L10.1|C20orf67|Pcif1|PCIF1_HUMAN|PDX1 C terminal inhibiting factor 1|Phosphorylated CTD interacting factor 1|Phosphorylated CTD-interacting factor 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H4Z3
- Q9H4Z3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together