Anti-PROK1 Rabbit Recombinant Polyclonal Antibody (HRP)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PROK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-497
- 77795-496
- Antigen Symbol:
- PROK1
- PROK1
- Antigen Name:
- Prokineticin-1 protein (20-105AA)
- Prokineticin-1 protein (20-105AA)
- Antigen Synonyms:
- Black mamba toxin related protein|EG VEGF|EG-VEGF|EGVEGF|Endocrine-gland-derived vascular endothelial growth factor|Mambakine|PK1|PRK1|PROK1|PROK1_HUMAN|Prokineticin 1|Prokineticin-1
- Black mamba toxin related protein|EG VEGF|EG-VEGF|EGVEGF|Endocrine-gland-derived vascular endothelial growth factor|Mambakine|PK1|PRK1|PROK1|PROK1_HUMAN|Prokineticin 1|Prokineticin-1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P58294
- P58294
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together