Anti-PUS10 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: PUS10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77820-381
- 77820-380
- Antigen Symbol:
- PUS10
- PUS10
- Antigen Name:
- Synthetic peptide of PUS10
- Synthetic peptide of PUS10
- Antigen Synonyms:
- CCDC139|Coiled coil domain containing 139|Coiled coil domain containing protein 139|Coiled-coil domain-containing protein 139|DOBI|FLJ32312|MGC126729|Pseudouridine synthase 10|Pseudouridylate synthase 10|Psi55 synthase|PUS10_HUMAN|Putative tRNA pseudouridine synthase Pus10|tRNA pseudouridine 55 synthase|tRNA pseudouridylate synthase|tRNA uridine isomerase|tRNA-uridine isomerase
- CCDC139|Coiled coil domain containing 139|Coiled coil domain containing protein 139|Coiled-coil domain-containing protein 139|DOBI|FLJ32312|MGC126729|Pseudouridine synthase 10|Pseudouridylate synthase 10|Psi55 synthase|PUS10_HUMAN|Putative tRNA pseudouridine synthase Pus10|tRNA pseudouridine 55 synthase|tRNA pseudouridylate synthase|tRNA uridine isomerase|tRNA-uridine isomerase
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q3MIT2
- Q3MIT2
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together