Anti-BCL2L15 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: BCL2L15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77820-660
- 77820-659
- Antigen Symbol:
- BCL2L15
- BCL2L15
- Antigen Name:
- Fusion protein of BCL2L15
- Fusion protein of BCL2L15
- Antigen Synonyms:
- AC124698.1|B2L15_HUMAN|Bcl-2-like protein 15|Bcl2-L-15|BCL2-like 15|Bfk|C1orf178|FLJ22588|Gm566|Pro apoptotic Bcl 2 protein
- AC124698.1|B2L15_HUMAN|Bcl-2-like protein 15|Bcl2-L-15|BCL2-like 15|Bfk|C1orf178|FLJ22588|Gm566|Pro apoptotic Bcl 2 protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5TBC7
- Q5TBC7
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together