To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Pre-mRNA-splicing regulator WTAP protein (1-151AA)
- Antigen Symbol:WTAP
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q15007
- Antigen Synonyms:DKFZp686F20131|Female-lethal(2)D homolog|FL2D_HUMAN|hFL(2)D|KIAA0105|MGC3925|Mum2|OTTHUMP00000017522|OTTHUMP00000017523|PNAS 132|PNAS-132|PNAS132|Pre mRNA splicing regulator WTAP|Pre-mRNA-splicing regulator WTAP|Putative pre mRNA splicing regulator female lethal 2D homolog|Putative pre mRNA splicing regulator female lethal(2D)|Putative pre-mRNA splicing regulator female lethal(2D) homolog|putative pre-mRNA splicing regulator female-lethal(2D)|Wilms tumor 1 associated protein|Wilms tumor 1 associating protein|Wilms tumor 1-associating protein|Wilms' tumor 1 associating protein|Wilms' tumour 1-associating protein|WT1 associated protein|WT1-associated protein|WT1-associating protein|wtap
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: WTAP
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat