Anti-SSTR4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:15 to 1:50
Type:
Antigen: SSTR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77819-928
- 77819-927
- Antigen Symbol:
- SSTR4
- SSTR4
- Antigen Name:
- Synthetic peptide of SSTR4
- Synthetic peptide of SSTR4
- Antigen Synonyms:
- Somatostatin Receptor 4|Somatostatin receptor type 4|SS 4R|SS-4-R|SS4-R|SS4R|SSR4_HUMAN|SST R4|SSTR 4|Sstr4
- Somatostatin Receptor 4|Somatostatin receptor type 4|SS 4R|SS-4-R|SS4-R|SS4R|SSR4_HUMAN|SST R4|SSTR 4|Sstr4
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31391
- P31391
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together