Anti-MOGAT2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: MOGAT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-212
- 77826-213
- Antigen Symbol:
- MOGAT2
- MOGAT2
- Antigen Name:
- 2-acylglycerol O-acyltransferase 2 protein (124-284AA)
- 2-acylglycerol O-acyltransferase 2 protein (124-284AA)
- Antigen Synonyms:
- 2 acylglycerol O acyltransferase 2|2-acylglycerol O-acyltransferase 2|Acyl CoA:monoacylglycerol acyltransferase 2|Acyl-CoA:monoacylglycerol acyltransferase 2|DC5|DGAT2L5|Diacylglycerol acyltransferase 2 like protein 5|Diacylglycerol acyltransferase 2-like protein 5|Diacylglycerol O acyltransferase candidate 5|Diacylglycerol O-acyltransferase candidate 5|EC 2.3.1.22|FLJ22644|hDC5|hMGAT2|Mgat1l|MGAT2|MGC119183|MGC119184|MGC119185|MGC189143|mogat2|MOGT2_HUMAN|Monoacylglycerol O acyltransferase 1 like|Monoacylglycerol O acyltransferase 2|Monoacylglycerol O-acyltransferase 2
- 2 acylglycerol O acyltransferase 2|2-acylglycerol O-acyltransferase 2|Acyl CoA:monoacylglycerol acyltransferase 2|Acyl-CoA:monoacylglycerol acyltransferase 2|DC5|DGAT2L5|Diacylglycerol acyltransferase 2 like protein 5|Diacylglycerol acyltransferase 2-like protein 5|Diacylglycerol O acyltransferase candidate 5|Diacylglycerol O-acyltransferase candidate 5|EC 2.3.1.22|FLJ22644|hDC5|hMGAT2|Mgat1l|MGAT2|MGC119183|MGC119184|MGC119185|MGC189143|mogat2|MOGT2_HUMAN|Monoacylglycerol O acyltransferase 1 like|Monoacylglycerol O acyltransferase 2|Monoacylglycerol O-acyltransferase 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q3SYC2
- Q3SYC2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together