Anti-RSPH3 Rabbit Polyclonal Antibody
77798-320
New Product
- Antigen Name:Radial spoke head-like protein 2
- Antigen Symbol:RSPH3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q86UC2
- Antigen Synonyms:RSPH3|RSHL2|RSP3|Radial spoke head protein 3 homolog|A-kinase anchor protein RSPH3|Radial spoke head-like protein 2
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Type:
Antigen: RSPH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat