Anti-PLXNA4 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PLXNA4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-745
- 77835-744
- Antigen Symbol:
- PLXNA4
- PLXNA4
- Antigen Name:
- Plexin-A4 protein (24-522AA)
- Plexin-A4 protein (24-522AA)
- Antigen Synonyms:
- FAYV2820|KIAA1550|PLEXA4|Plexin-A4|PlexinA4|PLXA4_HUMAN|plxna4|PLXNA4A|PLXNA4B|PRO34003|UNQ2820/PRO34003
- FAYV2820|KIAA1550|PLEXA4|Plexin-A4|PlexinA4|PLXA4_HUMAN|plxna4|PLXNA4A|PLXNA4B|PRO34003|UNQ2820/PRO34003
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HCM2
- Q9HCM2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together