Anti-SFRP2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SFRP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77799-324
- 77799-325
- Antigen Symbol:
- SFRP2
- SFRP2
- Antigen Name:
- Secreted frizzled-related protein 2 protein (68-186AA)
- Secreted frizzled-related protein 2 protein (68-186AA)
- Antigen Synonyms:
- AI851596|Frizzled related protein 2|FRP 2|FRP-2|FRP2|MGC128911|MGC153618|MGC53344|SARP 1|SARP-1|SARP1|SDF 5|SDF5|Secreted apoptosis related protein 1|Secreted apoptosis-related protein 1|Secreted frizzled related protein 2|Secreted frizzled-related protein 2|sFRP 2|sFRP-2|SFRP2|Sfrp2 protein|SFRP2_HUMAN
- AI851596|Frizzled related protein 2|FRP 2|FRP-2|FRP2|MGC128911|MGC153618|MGC53344|SARP 1|SARP-1|SARP1|SDF 5|SDF5|Secreted apoptosis related protein 1|Secreted apoptosis-related protein 1|Secreted frizzled related protein 2|Secreted frizzled-related protein 2|sFRP 2|sFRP-2|SFRP2|Sfrp2 protein|SFRP2_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96HF1
- Q96HF1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together