Anti-RSPH9 Rabbit Recombinant Polyclonal Antibody (HRP)
New Product
Type:
Antigen: RSPH9
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77837-097
- 77837-096
- Antigen Symbol:
- RSPH9
- RSPH9
- Antigen Name:
- Radial spoke head protein 9 homolog protein (62-171AA)
- Radial spoke head protein 9 homolog protein (62-171AA)
- Antigen Synonyms:
- C6orf206|Chromosome 6 open reading frame 206|CILD12|FLJ30845|Mitochondrial ribosomal protein S18A like 1|MRPS18AL1|Radial spoke head 9 homolog|Radial spoke head protein 9 homolog|RSPH 9|rsph9|RSPH9_HUMAN|Uncharacterized protein C6orf206
- C6orf206|Chromosome 6 open reading frame 206|CILD12|FLJ30845|Mitochondrial ribosomal protein S18A like 1|MRPS18AL1|Radial spoke head 9 homolog|Radial spoke head protein 9 homolog|RSPH 9|rsph9|RSPH9_HUMAN|Uncharacterized protein C6orf206
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9H1X1
- Q9H1X1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together