Anti-TK Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human herpesvirus 1
Specifications
- Catalog No:
- 77802-657
- 77802-656
- Antigen Symbol:
- TK
- TK
- Antigen Name:
- herpesvirus 1 Thymidine kinase protein (1-376AA)
- herpesvirus 1 Thymidine kinase protein (1-376AA)
- Antigen Synonyms:
- TK| UL23| Thymidine kinase| EC 2.7.1.21
- TK| UL23| Thymidine kinase| EC 2.7.1.21
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human herpesvirus 1
- Human herpesvirus 1
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P03176
- P03176
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together