About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:200 to 1:500, IF: 1:50 to 1:200
Type:
Antigen: ROCK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77822-397
- 77822-398
- Antigen Symbol:
- ROCK1
- ROCK1
- Antigen Name:
- Rho-associated protein kinase 1 protein (405-668AA)
- Rho-associated protein kinase 1 protein (405-668AA)
- Antigen Synonyms:
- coiled-coil-containing protein kinase 1|coiled-coil-containing protein kinase I|MGC131603|MGC43611|p160 Rhoassociated coiled coil-forming protein kinase|p160 ROCK-1|p160 p160ROCK|PRO0435|Renal carcinoma antigen NY REN 35|Renal carcinoma antigen NY-REN-35|Rho associated coiled coil containing protein kinase 1|Rho associated protein kinase 1|Rho kinase|Rho-alpha kinase|Rho-associated|Rho-associated protein kinase 1|ROCK I|ROCK-I|ROCK1_HUMAN|Rok|rokalpha
- coiled-coil-containing protein kinase 1|coiled-coil-containing protein kinase I|MGC131603|MGC43611|p160 Rhoassociated coiled coil-forming protein kinase|p160 ROCK-1|p160 p160ROCK|PRO0435|Renal carcinoma antigen NY REN 35|Renal carcinoma antigen NY-REN-35|Rho associated coiled coil containing protein kinase 1|Rho associated protein kinase 1|Rho kinase|Rho-alpha kinase|Rho-associated|Rho-associated protein kinase 1|ROCK I|ROCK-I|ROCK1_HUMAN|Rok|rokalpha
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13464
- Q13464
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice