Anti-KRT40 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: KRT40
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-124
- 77828-123
- Antigen Symbol:
- KRT40
- KRT40
- Antigen Name:
- Keratin, type I cytoskeletal 40 protein (1-200AA)
- Keratin, type I cytoskeletal 40 protein (1-200AA)
- Antigen Synonyms:
- CK-40|Cytokeratin-40|hair cortex Cytokeratin|K1C40_HUMAN|K40|Keratin|type I cytoskeletal 40|Keratin-40|Krt40|Type I hair keratin Ka36
- CK-40|Cytokeratin-40|hair cortex Cytokeratin|K1C40_HUMAN|K40|Keratin|type I cytoskeletal 40|Keratin-40|Krt40|Type I hair keratin Ka36
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q6A162
- Q6A162
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together