Anti-LRRK1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LRRK1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77826-028
- 77826-029
- Antigen Symbol:
- LRRK1
- LRRK1
- Antigen Name:
- Leucine-rich repeat serine/threonine-protein kinase 1 protein (1-68AA)
- Leucine-rich repeat serine/threonine-protein kinase 1 protein (1-68AA)
- Antigen Synonyms:
- KIAA1790|Leucine rich repeat serine/threonine protein kinase 1|Leucine-rich repeat serine/threonine-protein kinase 1|LRRK 1|Lrrk1|LRRK1_HUMAN|RIPK6|Roco1
- KIAA1790|Leucine rich repeat serine/threonine protein kinase 1|Leucine-rich repeat serine/threonine-protein kinase 1|LRRK 1|Lrrk1|LRRK1_HUMAN|RIPK6|Roco1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q38SD2
- Q38SD2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together