Anti-ATP6AP2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: ATP6AP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77813-989
- 77813-990
- Antigen Symbol:
- ATP6AP2
- ATP6AP2
- Antigen Name:
- Fusion protein of ATP6AP2
- Fusion protein of ATP6AP2
- Antigen Synonyms:
- APT6M8 9|APT6M8-9|ATP6IP2|ATP6M8-9|ATPase H(+)-transporting lysosomal accessory protein 2|ATPase H(+)-transporting lysosomal-interacting protein 2|ATPase H+ transporting lysosomal accessory protein 2|ATPase H+ transporting lysosomal interacting protein 2|ATPase H+ transporting lysosomal vacuolar proton pump membrane sector associated protein M8 9|ATPase membrane sector associated protein M8 9|ATPase|H+ transporting|lysosomal (vacuolar proton pump) membrane sector associated protein M8 9|CAPER|ELDF10|Embryonic liver differentiation factor 10|ER localized type I transmembrane adaptor|ER-localized type I transmembrane adaptor|HT028|M8 9|M8-9|MGC99577|MRXE|MSTP009|N14F|Renin receptor|Renin/prorenin receptor|RENR_HUMAN|V ATPase M8 9 subunit|V ATPase M8.9 subunit|V-ATPase M8.9 subunit|Vacuolar ATP synthase membrane sector associated protein M8 9|Vacuolar ATP synthase membrane sector-associated protein M8-9|vacuolar proton ATP synthase membrane sector associated protein M8 9|XMRE
- APT6M8 9|APT6M8-9|ATP6IP2|ATP6M8-9|ATPase H(+)-transporting lysosomal accessory protein 2|ATPase H(+)-transporting lysosomal-interacting protein 2|ATPase H+ transporting lysosomal accessory protein 2|ATPase H+ transporting lysosomal interacting protein 2|ATPase H+ transporting lysosomal vacuolar proton pump membrane sector associated protein M8 9|ATPase membrane sector associated protein M8 9|ATPase|H+ transporting|lysosomal (vacuolar proton pump) membrane sector associated protein M8 9|CAPER|ELDF10|Embryonic liver differentiation factor 10|ER localized type I transmembrane adaptor|ER-localized type I transmembrane adaptor|HT028|M8 9|M8-9|MGC99577|MRXE|MSTP009|N14F|Renin receptor|Renin/prorenin receptor|RENR_HUMAN|V ATPase M8 9 subunit|V ATPase M8.9 subunit|V-ATPase M8.9 subunit|Vacuolar ATP synthase membrane sector associated protein M8 9|Vacuolar ATP synthase membrane sector-associated protein M8-9|vacuolar proton ATP synthase membrane sector associated protein M8 9|XMRE
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O75787
- O75787
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together