Anti-CARD6 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CARD6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77835-910
- 77835-909
- Antigen Symbol:
- CARD6
- CARD6
- Antigen Name:
- Caspase recruitment domain-containing protein 6 protein (666-930AA)
- Caspase recruitment domain-containing protein 6 protein (666-930AA)
- Antigen Synonyms:
- CARD 6|CARD containing inhibitor of Nod1 and Cardiak-induced NF kB activation|CARD6|CARD6_HUMAN|Caspase recruitment domain family|member 6|Caspase recruitment domain protein 6|Caspase recruitment domain-containing protein 6|CINCIN1|D730008L15
- CARD 6|CARD containing inhibitor of Nod1 and Cardiak-induced NF kB activation|CARD6|CARD6_HUMAN|Caspase recruitment domain family|member 6|Caspase recruitment domain protein 6|Caspase recruitment domain-containing protein 6|CINCIN1|D730008L15
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BX69
- Q9BX69
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together