Anti-GFRA4 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: GFRA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77813-293
- 77813-292
- Antigen Symbol:
- GFRA4
- GFRA4
- Antigen Name:
- Synthetic peptide of GFRA4
- Synthetic peptide of GFRA4
- Antigen Synonyms:
- GDNF family receptor alpha 4|GDNF family receptor alpha-4|GDNF receptor alpha-4|GDNFR-alpha-4|GFR alpha 4|GFR receptor alpha 4|GFR-alpha-4|GFRA4|GFRA4_HUMAN|Glial cell line derived neurotrophic factor family receptor alpha 4|Neurotrophic factor receptor splice variant A (Gfra4 gene)|OTTHUMP00000030106|OTTHUMP00000030107|Persephin receptor
- GDNF family receptor alpha 4|GDNF family receptor alpha-4|GDNF receptor alpha-4|GDNFR-alpha-4|GFR alpha 4|GFR receptor alpha 4|GFR-alpha-4|GFRA4|GFRA4_HUMAN|Glial cell line derived neurotrophic factor family receptor alpha 4|Neurotrophic factor receptor splice variant A (Gfra4 gene)|OTTHUMP00000030106|OTTHUMP00000030107|Persephin receptor
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9GZZ7
- Q9GZZ7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together