Anti-UBXN6 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBXN6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77839-252
- 77839-251
- Antigen Symbol:
- UBXN6
- UBXN6
- Antigen Name:
- UBX domain-containing protein 6 protein (1-388AA)
- UBX domain-containing protein 6 protein (1-388AA)
- Antigen Synonyms:
- UBX domain containing 1|UBX domain containing 2|UBX domain containing protein 1|UBX domain containing protein 6|UBX domain protein 6|UBX domain-containing protein 1|UBX domain-containing protein 6|UBXD1|UBXDC2|Ubxn6|UBXN6_HUMAN
- UBX domain containing 1|UBX domain containing 2|UBX domain containing protein 1|UBX domain containing protein 6|UBX domain protein 6|UBX domain-containing protein 1|UBX domain-containing protein 6|UBXD1|UBXDC2|Ubxn6|UBXN6_HUMAN
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BZV1
- Q9BZV1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together