Anti-ATG3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: ATG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77832-869
- 77832-868
- Antigen Symbol:
- ATG3
- ATG3
- Antigen Name:
- Fusion protein of ATG3
- Fusion protein of ATG3
- Antigen Synonyms:
- 2610016C12Rik| Apg 3| APG3| APG3 autophagy 3 like| APG3 like| APG3, S. cerevisiae, homolog of| APG3-like| APG3L| Apg3p| ATG 3| ATG3| ATG3 autophagy related 3 homolog| ATG3 autophagy related 3 homolog (S. cerevisiae)| ATG3_HUMAN| Autophagy 3, S. cerevisiae, homolog of| Autophagy Apg3p/Aut1p like| autophagy related 3| Autophagy related protein 3| Autophagy-related protein 3| DKFZp564M1178| FLJ22125| hApg3| MGC15201| OTTHUMP00000214547| OTTHUMP00000214548| PC3 96| Protein PC3-96| Ubiquitin-like-conjugating enzyme ATG3
- 2610016C12Rik| Apg 3| APG3| APG3 autophagy 3 like| APG3 like| APG3, S. cerevisiae, homolog of| APG3-like| APG3L| Apg3p| ATG 3| ATG3| ATG3 autophagy related 3 homolog| ATG3 autophagy related 3 homolog (S. cerevisiae)| ATG3_HUMAN| Autophagy 3, S. cerevisiae, homolog of| Autophagy Apg3p/Aut1p like| autophagy related 3| Autophagy related protein 3| Autophagy-related protein 3| DKFZp564M1178| FLJ22125| hApg3| MGC15201| OTTHUMP00000214547| OTTHUMP00000214548| PC3 96| Protein PC3-96| Ubiquitin-like-conjugating enzyme ATG3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NT62
- Q9NT62
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together