Anti-cry1Ab Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: cry1Ab
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacillus thuringiensis subsp. kurstaki
Specifications
- Catalog No:
- 77817-651
- 77817-652
- Antigen Symbol:
- cry1Ab
- cry1Ab
- Antigen Name:
- Recombinant Bacillus thuringiensis subsp. kurstaki Pesticidal crystal protein Cry1Ab protein (1022-1155AA)
- Recombinant Bacillus thuringiensis subsp. kurstaki Pesticidal crystal protein Cry1Ab protein (1022-1155AA)
- Antigen Synonyms:
- cry1Ab|bt2|cry-1-2|cry1A(b)|cryIA(b)|cryIC1Pesticidal crystal protein Cry1Ab|130 kDa crystal protein|Crystaline entomocidal protoxin|Insecticidal delta-endotoxin CryIA(b)
- cry1Ab|bt2|cry-1-2|cry1A(b)|cryIA(b)|cryIC1Pesticidal crystal protein Cry1Ab|130 kDa crystal protein|Crystaline entomocidal protoxin|Insecticidal delta-endotoxin CryIA(b)
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Bacillus thuringiensis subsp. kurstaki
- Bacillus thuringiensis subsp. kurstaki
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P0A370
- P0A370
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, protein G purified
- >95%, protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together