Anti-TPM4 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: TPM4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-381
- 77803-381
- Antigen Symbol:
- TPM4
- TPM4
- Antigen Name:
- Tropomyosin alpha-4 chain protein (1-248AA)
- Tropomyosin alpha-4 chain protein (1-248AA)
- Antigen Synonyms:
- epididymis secretory protein Li 108|HEL-S-108|TM30p1|TMP 4|TPM4|TPM4_HUMAN|Tropomyosin 4|Tropomyosin alpha 4 chain|Tropomyosin alpha-4 chain|Tropomyosin-4|wu:fb06e07|zgc:103436|zgc:63909
- epididymis secretory protein Li 108|HEL-S-108|TM30p1|TMP 4|TPM4|TPM4_HUMAN|Tropomyosin 4|Tropomyosin alpha 4 chain|Tropomyosin alpha-4 chain|Tropomyosin-4|wu:fb06e07|zgc:103436|zgc:63909
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P67936
- P67936
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together