Anti-MYH14 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MYH14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77832-561
- 77832-560
- Antigen Symbol:
- MYH14
- MYH14
- Antigen Name:
- Myosin-14 protein (1058-1195AA)
- Myosin-14 protein (1058-1195AA)
- Antigen Synonyms:
- 2400004E04Rik| DFNA4| DKFZp667A1311| FLJ13881| FLJ43092| FP17425| II C| KIAA2034| MHC16| Myh 14| MYH14| MYH14_HUMAN| Myosin 14| Myosin| Myosin heavy chain 14| Myosin heavy chain| Myosin heavy chain non muscle IIc| Myosin heavy polypeptide 14| Myosin-14| NMHC II C| NMHC II-C| Non muscle myosin heavy chain IIc| non-muscle IIc| Non-muscle myosin heavy chain IIc| Nonmuscle myosin heavy chain II C| OTTMUSP00000019210
- 2400004E04Rik| DFNA4| DKFZp667A1311| FLJ13881| FLJ43092| FP17425| II C| KIAA2034| MHC16| Myh 14| MYH14| MYH14_HUMAN| Myosin 14| Myosin| Myosin heavy chain 14| Myosin heavy chain| Myosin heavy chain non muscle IIc| Myosin heavy polypeptide 14| Myosin-14| NMHC II C| NMHC II-C| Non muscle myosin heavy chain IIc| non-muscle IIc| Non-muscle myosin heavy chain IIc| Nonmuscle myosin heavy chain II C| OTTMUSP00000019210
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q7Z406
- Q7Z406
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together