Anti-TNFAIP1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TNFAIP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77824-805
- 77824-804
- Antigen Symbol:
- TNFAIP1
- TNFAIP1
- Antigen Name:
- BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2 protein (233-316AA)
- BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2 protein (233-316AA)
- Antigen Synonyms:
- TNFAIP1|BACURD2|EDP1BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2|hBACURD2|BTB/POZ domain-containing protein TNFAIP1|Protein B12|Tumor necrosis factor|alpha-induced protein 1|endothelial
- TNFAIP1|BACURD2|EDP1BTB/POZ domain-containing adapter for CUL3-mediated RhoA degradation protein 2|hBACURD2|BTB/POZ domain-containing protein TNFAIP1|Protein B12|Tumor necrosis factor|alpha-induced protein 1|endothelial
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13829
- Q13829
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together