Anti-ZNF384 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ZNF384
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77806-752
- 77806-753
- Antigen Symbol:
- ZNF384
- ZNF384
- Antigen Name:
- Zinc finger protein 384 protein (1-245AA)
- Zinc finger protein 384 protein (1-245AA)
- Antigen Synonyms:
- CAG repeat protein 1|CAGH1A|CAS interacting zinc finger protein|CAS-interacting zinc finger protein|CIZ|ERDA2|GH1A|NMP4|NP|Nuclear matrix protein 4|Nuclear matrix transcription factor 4|TNRC1|Trinucleotide repeat containing gene 1 protei|Trinucleotide repeat-containing gene 1 protein|Zinc finger protein 384|ZN384_HUMAN|Znf384
- CAG repeat protein 1|CAGH1A|CAS interacting zinc finger protein|CAS-interacting zinc finger protein|CIZ|ERDA2|GH1A|NMP4|NP|Nuclear matrix protein 4|Nuclear matrix transcription factor 4|TNRC1|Trinucleotide repeat containing gene 1 protei|Trinucleotide repeat-containing gene 1 protein|Zinc finger protein 384|ZN384_HUMAN|Znf384
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8TF68
- Q8TF68
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together