To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:Proteasome activator complex subunit 1 protein (1-249AA)
- Antigen Symbol:PSME1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q06323
- Antigen Synonyms:11S regulator complex alpha subunit|11S regulator complex subunit alpha|29 kD MCP activator subunit|29kD MCP activator subunit|Activator of multicatalytic protease subunit 1|IFI5111|IGUP I-5111|Interferon gamma IEF SSP 5111|Interferon gamma inducible protein 5111|Interferon gamma up regulated I 5111 protein|Interferon gamma up-regulated I-5111 protein|MGC8628|PA28a|PA28alpha|Proteasome (prosome, macropain) activator subunit 1 (PA28 alpha)|Proteasome activator 28 subunit alpha|Proteasome activator complex subunit 1|Proteasome activator subunit 1|PSME1|PSME1_HUMAN|REG-alpha|REGalpha
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200, IP:1:200 to 1:2000
Type:
Antigen: PSME1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse