Anti-SFT2D2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB: 1:1000 to 1:5000, IF: 1:50 to 1:200
Type:
Antigen: SFT2D2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77799-359
- 77799-358
- Antigen Symbol:
- SFT2D2
- SFT2D2
- Antigen Name:
- Vesicle transport protein SFT2B protein (1-36AA)
- Vesicle transport protein SFT2B protein (1-36AA)
- Antigen Synonyms:
- dJ747L4.C1.2|FLJ34085|SFT2 domain containing 2|SFT2 domain-containing protein 2|SFT2B_HUMAN|Sft2d2|UNQ512|Vesicle transport protein SFT2B
- dJ747L4.C1.2|FLJ34085|SFT2 domain containing 2|SFT2 domain-containing protein 2|SFT2B_HUMAN|Sft2d2|UNQ512|Vesicle transport protein SFT2B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O95562
- O95562
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together