Anti-EXD2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: EXD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77838-796
- 77838-795
- Antigen Symbol:
- EXD2
- EXD2
- Antigen Name:
- Exonuclease 3\\\'-5\\\' domain-containing protein 2 protein (137-245AA)
- Exonuclease 3\\\'-5\\\' domain-containing protein 2 protein (137-245AA)
- Antigen Synonyms:
- 4930539P14Rik| C14orf114| C85658| DKFZp781A0133| DKFZp781L15100| Exd2| EXD2_HUMAN| EXDL2| Exonuclease 3''-5'' domain-containing protein 2| Exonuclease 3''-5'' domain-like-containing protein 2| exonuclease 3'-5' domain-like 2| FLJ10738| RGD1311087
- 4930539P14Rik| C14orf114| C85658| DKFZp781A0133| DKFZp781L15100| Exd2| EXD2_HUMAN| EXDL2| Exonuclease 3''-5'' domain-containing protein 2| Exonuclease 3''-5'' domain-like-containing protein 2| exonuclease 3'-5' domain-like 2| FLJ10738| RGD1311087
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NVH0
- Q9NVH0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together