Anti-HLA-C Rabbit Recombinant Polyclonal Antibody
Catalog # 77815-216
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:HLA class I histocompatibility antigen, Cw-7 alpha chain protein (62-306AA)
- Antigen Symbol:HLA-C
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P10321
- Antigen Synonyms:1C07_HUMAN|Cw-7 alpha chain|D6S204|HLA class I histocompatibility antigen|C alpha chain|Cw-1 alpha chain|HLA JY3|HLA-C|HLA-C histocompatibility type|HLA-Cw|HLA-JY3|HLAC|HLC C|HLC-C|human leukocyte antigen-C alpha chain|major histocompatibility antigen HLA-C|major histocompatibility complex|class I|C|MHC class I antigen|MHC class I antigen Cw 1|MHC class I antigen Cw*7|MHC class I antigen heavy chain HLA-C|PSORS1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: HLA-C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse