Anti-PIK3R6 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIK3R6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77828-250
- 77828-251
- Antigen Symbol:
- PIK3R6
- PIK3R6
- Antigen Name:
- Phosphoinositide 3-kinase regulatory subunit 6 protein (566-667AA)
- Phosphoinositide 3-kinase regulatory subunit 6 protein (566-667AA)
- Antigen Synonyms:
- C17orf38|Chromosome 17 open reading frame 38|HsT41028|p84 PI3K adapter protein|p84 PIKAP|p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein|p87 PI3K adapter protein|p87PIKAP|Phosphatidylinositol 3-kinase|regulatory subunit 6|Phosphoinositide 3-kinase gamma adapter protein of 87 kDa|Phosphoinositide 3-kinase regulatory subunit 6|PI3Kgamma adapter protein of 87 kDa|PI3R6_HUMAN|PIK3R6
- C17orf38|Chromosome 17 open reading frame 38|HsT41028|p84 PI3K adapter protein|p84 PIKAP|p87 phosphoinositide 3-kinase gamma (PI3Kg) adapter protein|p87 PI3K adapter protein|p87PIKAP|Phosphatidylinositol 3-kinase|regulatory subunit 6|Phosphoinositide 3-kinase gamma adapter protein of 87 kDa|Phosphoinositide 3-kinase regulatory subunit 6|PI3Kgamma adapter protein of 87 kDa|PI3R6_HUMAN|PIK3R6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q5UE93
- Q5UE93
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together