Anti-PRMT7 Rabbit Recombinant Polyclonal Antibody
77842-443
New Product
- Antigen Name:Protein arginine N-methyltransferase 7 protein (285-390AA)
- Antigen Symbol:PRMT7
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NVM4
- Antigen Synonyms:[Myelin basic protein]-arginine N-methyltransferase PRMT7|ANM7_HUMAN|FLJ10640|Histone-arginine N-methyltransferase PRMT7|KIAA1933|Myelin basic protein arginine N methyltransferase|OTTHUMP00000174863|PRMT7|Protein arginine methyltransferase 7|Protein arginine N-methyltransferase 7
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: PRMT7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse