Anti-LPAR4 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:25 to 1:100
Type:
Antigen: LPAR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77806-990
- 77806-989
- Antigen Symbol:
- LPAR4
- LPAR4
- Antigen Name:
- Synthetic peptide of LPAR4
- Synthetic peptide of LPAR4
- Antigen Synonyms:
- 5730485F04Rik|G protein coupled receptor 23|G-protein coupled receptor 23|GPR23|LPA receptor 4|LPA-4|LPA4|Lpar4|LPAR4_HUMAN|Lysophosphatidic acid receptor 4|Lysophosphatidic Acid Receptor P2Y9|P2RY9|P2Y purinoceptor 9|P2Y5 like|P2Y5-like receptor|P2Y9|Purinergic receptor 9|Purinergic receptor, P2Y, G-protein coupled, 9
- 5730485F04Rik|G protein coupled receptor 23|G-protein coupled receptor 23|GPR23|LPA receptor 4|LPA-4|LPA4|Lpar4|LPAR4_HUMAN|Lysophosphatidic acid receptor 4|Lysophosphatidic Acid Receptor P2Y9|P2RY9|P2Y purinoceptor 9|P2Y5 like|P2Y5-like receptor|P2Y9|Purinergic receptor 9|Purinergic receptor, P2Y, G-protein coupled, 9
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99677
- Q99677
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% Sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together