Anti-PIAS2 Rabbit Recombinant Polyclonal Antibody
Catalog # 77794-096
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:E3 SUMO-protein ligase PIAS2 protein (532-621AA)
- Antigen Symbol:PIAS2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O75928
- Antigen Synonyms:Androgen receptor interacting protein 3|Androgen receptor-interacting protein 3|ARIP3|DAB2 interacting protein|DAB2-interacting protein|DIP|E3 SUMO protein ligase PIAS2|E3 SUMO-protein ligase PIAS2|MIZ|Miz1|Msx interacting zinc finger protein|Msx-interacting zinc finger protein|PIAS NY protein|PIAS-NY protein|PIAS2|PIAS2_HUMAN|PIASX|PIASX-ALPHA|PIASX-BETA|Protein inhibitor of activated STAT x|Protein inhibitor of activated STAT, 2|Protein inhibitor of activated STAT2|SIZ2|Zinc finger, MIZ-type containing 4|ZMIZ4
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000
Type:
Antigen: PIAS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse