Anti-STAC Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: STAC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77834-697
- 77834-698
- Antigen Symbol:
- STAC
- STAC
- Antigen Name:
- SH3 and cysteine-rich domain-containing protein (1-119AA)
- SH3 and cysteine-rich domain-containing protein (1-119AA)
- Antigen Synonyms:
- FLJ32331|SH3 and cysteine rich domain|SH3 and cysteine rich domain containing protein|SH3 and cysteine-rich domain-containing protein|SRC homology 3 and cysteine rich domain protein|Src homology 3 and cysteine-rich domain-containing protein|Src homology three (SH3) and cysteine rich domain|STAC 1|STAC|STAC_HUMAN|STAC1
- FLJ32331|SH3 and cysteine rich domain|SH3 and cysteine rich domain containing protein|SH3 and cysteine-rich domain-containing protein|SRC homology 3 and cysteine rich domain protein|Src homology 3 and cysteine-rich domain-containing protein|Src homology three (SH3) and cysteine rich domain|STAC 1|STAC|STAC_HUMAN|STAC1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99469
- Q99469
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Upon receipt, store at −20...−80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together