Anti-PIP4P2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PIP4P2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-087
- 77803-087
- Antigen Symbol:
- PIP4P2
- PIP4P2
- Antigen Name:
- Type 2 phosphatidylinositol 4,5-bisphosphate 4-phosphatase protein (1-191AA)
- Type 2 phosphatidylinositol 4,5-bisphosphate 4-phosphatase protein (1-191AA)
- Antigen Synonyms:
- 2610319K07Rik|5-bisphosphate 4-phosphatase|5-P2 4-Ptase II|AI315591|AV001360|PtdIns-4|RGD1306225|RP23-379L17.2|TM55A_HUMAN|tmem55a|Transmembrane protein 55A|Type II phosphatidylinositol 4
- 2610319K07Rik|5-bisphosphate 4-phosphatase|5-P2 4-Ptase II|AI315591|AV001360|PtdIns-4|RGD1306225|RP23-379L17.2|TM55A_HUMAN|tmem55a|Transmembrane protein 55A|Type II phosphatidylinositol 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N4L2
- Q8N4L2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together