Anti-ROR2 Rabbit Polyclonal Antibody
Catalog # 77797-865
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from the Internal region of ROR2
- Antigen Symbol:ROR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q01974
- Antigen Synonyms:BDB|BDB1|Brachydactyly type B|EC 2.7.10.1|MGC163394|Neurotrophic tyrosine kinase|Neurotrophic tyrosine kinase, receptor related 2|NTRKR2|Receptor tyrosine kinase-like orphan receptor 2|receptor-related 2|ROR2|ROR2_HUMAN|Tyrosine protein kinase transmembrane receptor ROR2|Tyrosine-protein kinase transmembrane receptor Ror2
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, ELISA: 1:5000
Type:
Antigen: ROR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat