Anti-SULT1A1 Rabbit Recombinant Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SULT1A1
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77801-539
- 77801-538
- Antigen Symbol:
- SULT1A1
- SULT1A1
- Antigen Name:
- Sulfotransferase 1A1 protein (1-295AA)
- Sulfotransferase 1A1 protein (1-295AA)
- Antigen Synonyms:
- Aryl sulfotransferase 1|HAST1/HAST2|Human aryl sulfotransferase mRNA complete cds|MGC131921|MGC5163|OTTHUMP00000162568|OTTHUMP00000162569|P PST 1|P PST|P-PST 1|Phenol sulfating phenol sulfotransferase 1|Phenol sulfotransferase 1|Phenol-sulfating phenol sulfotransferase 1|PST|ST1A1|ST1A1_HUMAN|ST1A3|STP|STP1|Sulfotransferase 1A1|Sulfotransferase family 1A phenol preferring member 1|Sulfotransferase family cytosolic 1A phenol preferring member 1|Sulfotransferase phenol preferring 1|SULT1A1|Thermostable phenol sulfotransferase|Thermostable phenol sulfotransferase1|Ts-PST|TSPST1
- Aryl sulfotransferase 1|HAST1/HAST2|Human aryl sulfotransferase mRNA complete cds|MGC131921|MGC5163|OTTHUMP00000162568|OTTHUMP00000162569|P PST 1|P PST|P-PST 1|Phenol sulfating phenol sulfotransferase 1|Phenol sulfotransferase 1|Phenol-sulfating phenol sulfotransferase 1|PST|ST1A1|ST1A1_HUMAN|ST1A3|STP|STP1|Sulfotransferase 1A1|Sulfotransferase family 1A phenol preferring member 1|Sulfotransferase family cytosolic 1A phenol preferring member 1|Sulfotransferase phenol preferring 1|SULT1A1|Thermostable phenol sulfotransferase|Thermostable phenol sulfotransferase1|Ts-PST|TSPST1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P50225
- P50225
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together