Anti-CHD5 Rabbit Polyclonal Antibody
Catalog # 77813-128
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthetic peptide corresponding to residues near the C terminal of Cadherin 5, type 2 (vascular endothelium)
- Antigen Symbol:CHD5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Size:100 µl
- Form:Liquid
- Gene ID:Q8TDI0
- Antigen Synonyms:ATP-dependent helicase CHD5|CHD 5|CHD-5|CHD5|CHD5_HUMAN|Chromodomain-helicase-DNA-binding protein 5|DKFZp434N231|KIAA0444|RP1-233K16.2
- Storage Buffer:PBS, 0.05% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:50 to 1:100
Type:
Antigen: CHD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat