Anti-PSMA3 Rabbit Recombinant Polyclonal Antibody (FITC)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: PSMA3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77795-763
- 77795-764
- Antigen Symbol:
- PSMA3
- PSMA3
- Antigen Name:
- Proteasome subunit alpha type-3 protein (218-248AA)
- Proteasome subunit alpha type-3 protein (218-248AA)
- Antigen Synonyms:
- HC8|Macropain subunit C8|MGC12306|MGC32631|Multicatalytic endopeptidase complex subunit C8|Proteasome (prosome macropain) subunit alpha type 3|Proteasome alpha 3 subunit|Proteasome component C8|Proteasome subunit alpha type 3|Proteasome subunit alpha type-3|Proteasome subunit C8|PSA3_HUMAN|PSC8|psmA3
- HC8|Macropain subunit C8|MGC12306|MGC32631|Multicatalytic endopeptidase complex subunit C8|Proteasome (prosome macropain) subunit alpha type 3|Proteasome alpha 3 subunit|Proteasome component C8|Proteasome subunit alpha type 3|Proteasome subunit alpha type-3|Proteasome subunit C8|PSA3_HUMAN|PSC8|psmA3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P25788
- P25788
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together