Anti-CDK17 Rabbit Recombinant Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CDK17
Clonality: Recombinant
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77813-813
- 77813-812
- Antigen Symbol:
- CDK17
- CDK17
- Antigen Name:
- Cyclin-dependent kinase 17 protein (337-523AA)
- Cyclin-dependent kinase 17 protein (337-523AA)
- Antigen Synonyms:
- 6430598J10Rik|CDK17|CDK17_HUMAN|Cell division protein kinase 17|Cyclin-dependent kinase 17|EC 2.7.1.37|MGC25109|PCTAIRE-motif protein kinase 2|PCTK2|Protein kinase cdc2-related PCTAIRE-2|Serine/threonine-protein kinase PCTAIRE-2
- 6430598J10Rik|CDK17|CDK17_HUMAN|Cell division protein kinase 17|Cyclin-dependent kinase 17|EC 2.7.1.37|MGC25109|PCTAIRE-motif protein kinase 2|PCTK2|Protein kinase cdc2-related PCTAIRE-2|Serine/threonine-protein kinase PCTAIRE-2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q00537
- Q00537
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together