Anti-UBE2D1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: UBE2D1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77811-836
- 77803-836
- Antigen Symbol:
- UBE2D1
- UBE2D1
- Antigen Name:
- Ubiquitin-conjugating enzyme E2 D1 protein (1-147AA)
- Ubiquitin-conjugating enzyme E2 D1 protein (1-147AA)
- Antigen Synonyms:
- E2(17)KB1|SFT|Stimulator of Fe transport|UB2D1_HUMAN|UBC 4/5|UBC4/5|UBC4/5 homolog|UBC4/5, S. cerevisiae, homolog of|UBCH 5|UBCH 5A|UbcH5|UBCH5A|Ube2d1|Ubiquitin carrier protein|Ubiquitin carrier protein D1|Ubiquitin conjugating enzyme E2 17 kDa 1|Ubiquitin conjugating enzyme E2 D1|Ubiquitin conjugating enzyme E2D 1 (UBC4/5 homolog yeast)|Ubiquitin conjugating enzyme E2D 1|Ubiquitin conjugating enzyme UBCH5A|Ubiquitin protein ligase D1|Ubiquitin-conjugating enzyme E2 D1|Ubiquitin-conjugating enzyme E2(17)KB 1|Ubiquitin-conjugating enzyme E2-17 kDa 1|Ubiquitin-protein ligase D1
- E2(17)KB1|SFT|Stimulator of Fe transport|UB2D1_HUMAN|UBC 4/5|UBC4/5|UBC4/5 homolog|UBC4/5, S. cerevisiae, homolog of|UBCH 5|UBCH 5A|UbcH5|UBCH5A|Ube2d1|Ubiquitin carrier protein|Ubiquitin carrier protein D1|Ubiquitin conjugating enzyme E2 17 kDa 1|Ubiquitin conjugating enzyme E2 D1|Ubiquitin conjugating enzyme E2D 1 (UBC4/5 homolog yeast)|Ubiquitin conjugating enzyme E2D 1|Ubiquitin conjugating enzyme UBCH5A|Ubiquitin protein ligase D1|Ubiquitin-conjugating enzyme E2 D1|Ubiquitin-conjugating enzyme E2(17)KB 1|Ubiquitin-conjugating enzyme E2-17 kDa 1|Ubiquitin-protein ligase D1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P51668
- P51668
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together