Anti-ORAOV1 Rabbit Polyclonal Antibody
Catalog # 77818-631
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Name:Synthesized peptide derived from internal of ORAV1.
- Antigen Symbol:ORAOV1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q8WV07
- Antigen Synonyms:LTO1|ORAOV1|TAOS1Protein LTO1 homolog|Oral cancer-overexpressed protein 1|Tumor-amplified and overexpressed sequence 1
- Storage Buffer:PBS, 150mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: ORAOV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human